siRNA Details
| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081075.1 | NP_001074544.1 | 3, 4 | 337 | ||
| GenBank mRNA | AK017666.1 | - | 3, 4 | 399 | ||
| AK077572.1 | - | 3, 4 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fra10ac1| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081075.1 | NP_001074544.1 | 3, 4 | 337 | ||
| GenBank mRNA | AK017666.1 | - | 3, 4 | 399 | ||
| AK077572.1 | - | 3, 4 | 310 | |||