siRNA Details
| Gene Symbol: 5730455P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027472.3 | NP_081748.1 | 9 | 975 | ||
| GenBank mRNA | AK017667.1 | - | 9 | 966 | ||
| AK078548.1 | - | 9 | 975 | |||
| AK135352.1 | - | 9 | 957 | |||
| AK153246.1 | - | 9 | 974 | |||
| AK160515.1 | - | 7 | 618 | |||
| BC044897.1 | - | 9 | 963 | 3990240 | ||
| BC055024.1 | - | 9 | 959 | 5149944 | ||

5730455P16Rik
Genomic Map