siRNA Details
| Gene Symbol: 5730455P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027472.3 | NP_081748.1 | 5 | 566 | ||
| GenBank mRNA | AK017667.1 | - | 5 | 557 | ||
| AK078548.1 | - | 5 | 567 | |||
| AK135352.1 | - | 5 | 548 | |||
| AK153246.1 | - | 5 | 565 | |||
| AK160515.1 | - | 3 | 209 | |||
| BC044897.1 | - | 5 | 554 | 3990240 | ||
| BC055024.1 | - | 5 | 550 | 5149944 | ||

5730455P16Rik
Genomic Map