siRNA Details
| Gene Symbol: Dnajb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033155.1 | NP_001028327.1 | 7 | 1132 | ||
| GenBank mRNA | AK017735.1 | - | 7 | 993 | ||
| AK135329.1 | - | 1 | 696 | |||
| BC117717.1 | - | 7 | 933 | 40088109 | ||
| BC117718.1 | - | 7 | 933 | 40088110 | ||

Dnajb14
Genomic Map