siRNA Details
| Gene Symbol: Fbxo33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033156.4 | NP_001028328.2 | 4 | 3014 | ||
| GenBank mRNA | AK017744.1 | - | 1 | 934 | ||
| AK132233.1 | - | 2 | 2973 | |||
| BC006669.1 | - | 2 | 2147 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108023, XM_006240119.

Fbxo33
Genomic Map