siRNA Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081430.1 | NP_001074899.1 | 5 | 2601 | ||
| GenBank mRNA | AK017805.1 | - | 1 | 412 | ||
| AK044235.1 | - | 4 | 2221 | |||
| BC054060.1 | - | 3 | 1828 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naa30| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081430.1 | NP_001074899.1 | 5 | 2601 | ||
| GenBank mRNA | AK017805.1 | - | 1 | 412 | ||
| AK044235.1 | - | 4 | 2221 | |||
| BC054060.1 | - | 3 | 1828 | |||