siRNA Details
| Gene Symbol: Utp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175158.3 | NP_780367.2 | 12, 13 | 1601 | ||
| GenBank mRNA | AK145554.1 | - | 12, 13 | 1570 | ||
| BC084586.1 | - | 12, 13 | 1600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utp20| Gene Symbol: Utp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175158.3 | NP_780367.2 | 12, 13 | 1601 | ||
| GenBank mRNA | AK145554.1 | - | 12, 13 | 1570 | ||
| BC084586.1 | - | 12, 13 | 1600 | |||