siRNA Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133911.1 | NP_598672.1 | 3, 4 | 598 | ||
| GenBank mRNA | AK014455.1 | - | 3, 4 | 570 | ||
| AK028929.1 | - | 3, 4 | 537 | |||
| BC052391.1 | - | 3, 4 | 598 | |||
More Information
Set Membership: |
|

Adgra2
Genomic Map