siRNA Details
| Gene Symbol: 6330409D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027529.1 | NP_081805.1 | 1 | 342 | ||
| GenBank mRNA | AK018150.1 | - | 1 | 344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330409D20Rik| Gene Symbol: 6330409D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027529.1 | NP_081805.1 | 1 | 342 | ||
| GenBank mRNA | AK018150.1 | - | 1 | 344 | ||