siRNA Details
Gene Symbol: Cep192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027556.1 | NP_081832.1 | 10 | 1253 | ||
GenBank mRNA | AK052420.1 | - | 10 | 1255 | ||
AK158414.1 | - | 10 | 1228 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cep192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027556.1 | NP_081832.1 | 10 | 1253 | ||
GenBank mRNA | AK052420.1 | - | 10 | 1255 | ||
AK158414.1 | - | 10 | 1228 |