siRNA Details
| Gene Symbol: Cep192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027556.1 | NP_081832.1 | 42 | 7317 | ||
| GenBank mRNA | AK014527.1 | - | 14 | 1916 | ||
| AK122526.1 | - | 22 | 3774 | |||
| BC024918.1 | - | 14 | 1874 | |||
| BC055776.1 | - | 14 | 1831 | |||
| BC064462.1 | - | 23 | 3359 | |||

Cep192
Genomic Map