siRNA Details
| Gene Symbol: Fam47c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164739.1 | NP_001158211.1 | 1 | 569 | ||
| GenBank mRNA | AK014848.1 | - | 1 | 570 | ||
| BC115971.1 | - | 1 | 540 | 40106467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam47c| Gene Symbol: Fam47c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164739.1 | NP_001158211.1 | 1 | 569 | ||
| GenBank mRNA | AK014848.1 | - | 1 | 570 | ||
| BC115971.1 | - | 1 | 540 | 40106467 | ||