siRNA Details
| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027584.1 | NP_081860.1 | 12, 13 | 2042 | ||
| GenBank mRNA | AK014872.1 | - | 12, 13 | 2043 | ||
| AK133206.1 | - | 13, 14 | 2131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco6d1| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027584.1 | NP_081860.1 | 12, 13 | 2042 | ||
| GenBank mRNA | AK014872.1 | - | 12, 13 | 2043 | ||
| AK133206.1 | - | 13, 14 | 2131 | |||