siRNA Details
| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164233.1 | NP_001157705.1 | 6, 7 | 1147 | ||
| NM_027584.1 | NP_081860.1 | 6, 7 | 1147 | |||
| GenBank mRNA | AK014872.1 | - | 6, 7 | 1148 | ||
| BC112423.1 | - | 6, 7 | 1117 | 40062055 | ||
| BC113131.1 | - | 6, 7 | 1117 | 40062053 | ||

Slco6d1
Genomic Map