siRNA Details
| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027585.2 | NP_081861.2 | 3 | 469 | ||
| GenBank mRNA | AK014912.1 | - | 3 | 492 | ||
| AK162372.1 | - | 3 | 350 | |||
| BC027204.1 | - | 3 | 309 | 4037010 | ||
| BC140978.1 | - | 3 | 452 | 9055590 | ||
| JN629039.1 | - | 3 | 409 | |||

Cnbd2
Genomic Map