siRNA Details
| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027598.1 | NP_081874.1 | 10 | 1422 | ||
| GenBank mRNA | AK014961.1 | - | 10 | 1423 | ||
| AK161257.1 | - | 804 | ||||
| BC049627.1 | - | 8 | 1267 | 6706381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921524L21Rik| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027598.1 | NP_081874.1 | 10 | 1422 | ||
| GenBank mRNA | AK014961.1 | - | 10 | 1423 | ||
| AK161257.1 | - | 804 | ||||
| BC049627.1 | - | 8 | 1267 | 6706381 | ||