siRNA Details
| Gene Symbol: Mmrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163507.1 | NP_001156979.1 | 6 | 2788 | ||
| NM_027613.1 | NP_081889.1 | 6 | 2788 | |||
| GenBank mRNA | AK036724.1 | - | 6 | 2627 | ||
| AK137556.1 | - | 6 | 2628 | |||
| BC046425.1 | - | 1 | 88 | |||
| BC137623.1 | - | 6 | 2588 | 8860642 | ||
| BC144789.1 | - | 6 | 2588 | 9053337 | ||

Mmrn1
Genomic Map