siRNA Details
| Gene Symbol: Ccdc105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027630.1 | NP_081906.1 | 6 | 1292 | ||
| GenBank mRNA | AK016452.1 | - | 6 | 1293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc105| Gene Symbol: Ccdc105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027630.1 | NP_081906.1 | 6 | 1292 | ||
| GenBank mRNA | AK016452.1 | - | 6 | 1293 | ||