siRNA Details
| Gene Symbol: Spef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027641.2 | NP_081917.1 | 7 | 1439 | ||
| GenBank mRNA | AK016476.1 | - | 7 | 1459 | ||
| AK046964.1 | - | 1505 | ||||
| AK047675.1 | - | 7 | 1440 | |||
| AY860964.1 | - | 7 | 1457 | |||
| BC006046.1 | - | 7 | 1379 | 3590641 | ||

Spef1
Genomic Map