siRNA Details
| Gene Symbol: Spef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027641.2 | NP_081917.1 | 6, 7 | 732 | ||
| GenBank mRNA | AK016476.1 | - | 6, 7 | 752 | ||
| AK046964.1 | - | 798 | ||||
| AK047675.1 | - | 6, 7 | 733 | |||
| AY860964.1 | - | 6, 7 | 750 | |||
| BC006046.1 | - | 6, 7 | 672 | 3590641 | ||

Spef1
Genomic Map