siRNA Details
| Gene Symbol: D7Ertd443e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199941.1 | NP_001186870.1 | 1 | 97 | ||
| GenBank mRNA | GQ499374.1 | - | 1 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D7Ertd443e| Gene Symbol: D7Ertd443e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199941.1 | NP_001186870.1 | 1 | 97 | ||
| GenBank mRNA | GQ499374.1 | - | 1 | 97 | ||