siRNA Details
| Gene Symbol: 4933403O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177389.1 | NP_001170860.1 | 3 | 1204 | ||
| GenBank mRNA | AK016644.1 | - | 3 | 1201 | ||
| BC115851.1 | - | 3 | 756 | |||
| BC127170.1 | - | 2 | 1092 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933403O08Rik| Gene Symbol: 4933403O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177389.1 | NP_001170860.1 | 3 | 1204 | ||
| GenBank mRNA | AK016644.1 | - | 3 | 1201 | ||
| BC115851.1 | - | 3 | 756 | |||
| BC127170.1 | - | 2 | 1092 | |||