siRNA Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019214.1 | NP_062087.1 | 2, 3 | 333 | ||
| XM_008764576.1 | XP_008762798.1 | 3, 4 | 1657 | |||
| GenBank mRNA | AF167412.1 | - | 2, 3 | 333 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a4| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019214.1 | NP_062087.1 | 2, 3 | 333 | ||
| XM_008764576.1 | XP_008762798.1 | 3, 4 | 1657 | |||
| GenBank mRNA | AF167412.1 | - | 2, 3 | 333 | ||