siRNA Details
| Gene Symbol: Prss55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081063.1 | NP_001074532.1 | 3 | 421 | ||
| GenBank mRNA | AK016601.1 | - | 2 | 455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss55| Gene Symbol: Prss55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081063.1 | NP_001074532.1 | 3 | 421 | ||
| GenBank mRNA | AK016601.1 | - | 2 | 455 | ||