siRNA Details
| Gene Symbol: 4933405L10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027655.1 | NP_081931.1 | 1 | 18 | ||
| GenBank mRNA | AK016673.1 | - | 1 | 31 | ||
| BC125412.1 | - | 1 | 18 | 40129927 | ||
| BC138007.1 | - | 1 | 18 | 8861026 | ||
| BC145093.1 | - | 1 | 18 | 9053641 | ||

4933405L10Rik
Genomic Map