siRNA Details
| Gene Symbol: Neurod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019218.2 | NP_062091.1 | 2 | 212 | ||
| GenBank mRNA | AF107728.1 | - | 1 | 125 | ||
| BC092367.1 | - | 2 | 212 | 7312547 | ||
| BC094526.1 | - | 1 | 429 | 7318277 | ||
| D82074.1 | - | 2 | 222 | |||
| D82075.1 | - | 1 | 1689 | |||
| D82945.1 | - | 1 | 125 | |||

Neurod1
Genomic Map