siRNA Details
| Gene Symbol: 4933412E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027668.1 | NP_081944.1 | 1 | 1136 | ||
| GenBank mRNA | AK016789.1 | - | 1 | 1132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933412E24Rik| Gene Symbol: 4933412E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027668.1 | NP_081944.1 | 1 | 1136 | ||
| GenBank mRNA | AK016789.1 | - | 1 | 1132 | ||