siRNA Details
| Gene Symbol: Glt6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039095.1 | NP_001034184.1 | 5 | 995 | ||
| GenBank mRNA | AK016765.1 | - | 5 | 1002 | ||
| BC111823.1 | - | 5 | 995 | 40052059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glt6d1| Gene Symbol: Glt6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039095.1 | NP_001034184.1 | 5 | 995 | ||
| GenBank mRNA | AK016765.1 | - | 5 | 1002 | ||
| BC111823.1 | - | 5 | 995 | 40052059 | ||