siRNA Details
| Gene Symbol: Tmem217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162901.1 | NP_001156373.1 | 3 | 277 | ||
| NM_001162902.1 | NP_001156374.1 | 2 | 185 | |||
| GenBank mRNA | AK016810.1 | - | 3 | 278 | ||
| BC116865.1 | - | 2 | 149 | 40126184 | ||
| BC116867.1 | - | 3 | 241 | 40126186 | ||
| BC144829.1 | - | 2 | 149 | 9053377 | ||

Tmem217
Genomic Map