siRNA Details
Gene Symbol: 4933416I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027700.1 | NP_081976.1 | 1219 | |||
GenBank mRNA | AK016833.1 | - | 1186 | |||
AK163858.1 | - | 1035 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933416I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027700.1 | NP_081976.1 | 1219 | |||
GenBank mRNA | AK016833.1 | - | 1186 | |||
AK163858.1 | - | 1035 |