siRNA Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027704.2 | NP_081980.1 | 1 | 50 | ||
| GenBank mRNA | AK016865.1 | - | 1 | 62 | ||
| BC116803.1 | - | 1 | 50 | 40126122 | ||
| BC116807.1 | - | 1 | 50 | 40126126 | ||
| BC144831.1 | - | 1 | 50 | |||
| BC144832.1 | - | 1 | 50 | |||

Zdhhc11
Genomic Map