siRNA Details
| Gene Symbol: Fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027708.1 | NP_081984.1 | 8 | 1339 | ||
| GenBank mRNA | AK016870.1 | - | 8 | 1340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo24| Gene Symbol: Fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027708.1 | NP_081984.1 | 8 | 1339 | ||
| GenBank mRNA | AK016870.1 | - | 8 | 1340 | ||