siRNA Details
Gene Symbol: Gpr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162955.1 | NP_001156427.1 | 1 | 535 | ||
GenBank mRNA | BC107229.2 | - | 1 | 535 | 40052032 | |
BC107281.2 | - | 1 | 533 | |||
BC107282.2 | - | 1 | 533 | |||
BC119501.1 | - | 1 | 535 | 40052770 | ||
BC119502.1 | - | 1 | 535 | 40052772 | ||
BC119518.1 | - | 1 | 535 | 40055788 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
