siRNA Details
Gene Symbol: Rnf148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027754.1 | NP_082030.1 | 1 | 793 | ||
GenBank mRNA | AK017027.1 | - | 1 | 794 | ||
AY155440.1 | - | 1 | 745 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027754.1 | NP_082030.1 | 1 | 793 | ||
GenBank mRNA | AK017027.1 | - | 1 | 794 | ||
AY155440.1 | - | 1 | 745 |