siRNA Details
Gene Symbol: Krtap7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027771.1 | NP_082047.1 | 1 | 537 | ||
GenBank mRNA | AK017387.1 | - | 1 | 538 | ||
CB320982.1 | - | 1 | 520 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027771.1 | NP_082047.1 | 1 | 537 | ||
GenBank mRNA | AK017387.1 | - | 1 | 538 | ||
CB320982.1 | - | 1 | 520 |