siRNA Details
| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019231.2 | NP_062104.2 | 4 | 452 | ||
| GenBank mRNA | AF092534.1 | - | 4 | 394 | ||
| BC107849.1 | - | 4 | 452 | 7936049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapk13| Gene Symbol: Mapk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019231.2 | NP_062104.2 | 4 | 452 | ||
| GenBank mRNA | AF092534.1 | - | 4 | 394 | ||
| BC107849.1 | - | 4 | 452 | 7936049 | ||