siRNA Details
Gene Symbol: Fam83c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027788.2 | NP_082064.2 | 1 | 538 | ||
GenBank mRNA | BC113190.1 | - | 1 | 537 | 40062623 | |
BC113797.1 | - | 1 | 537 | 40062621 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam83c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027788.2 | NP_082064.2 | 1 | 538 | ||
GenBank mRNA | BC113190.1 | - | 1 | 537 | 40062623 | |
BC113797.1 | - | 1 | 537 | 40062621 |