siRNA Details
| Gene Symbol: Fam83c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027788.2 | NP_082064.2 | 3 | 865 | ||
| GenBank mRNA | BC113190.1 | - | 3 | 864 | 40062623 | |
| BC113797.1 | - | 3 | 864 | 40062621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83c| Gene Symbol: Fam83c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027788.2 | NP_082064.2 | 3 | 865 | ||
| GenBank mRNA | BC113190.1 | - | 3 | 864 | 40062623 | |
| BC113797.1 | - | 3 | 864 | 40062621 | ||