siRNA Details
| Gene Symbol: Isx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294278.1 | NP_001281207.1 | 2 | 775 | ||
| NM_027837.3 | NP_082113.2 | 2 | 775 | |||
| GenBank mRNA | AB219124.1 | - | 2 | 308 | ||
| AK018615.1 | - | 2 | 776 | |||
| BC128289.1 | - | 2 | 409 | 40102442 | ||
| BC128290.1 | - | 2 | 409 | 40102443 | ||

Isx
Genomic Map