siRNA Details
Gene Symbol: Zfp949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142943.1 | NP_001136415.1 | 6 | 1531 | ||
NM_001163061.1 | NP_001156533.1 | 5 | 1385 | |||
GenBank mRNA | BC141009.1 | - | 6 | 1368 | 9055621 | |
BC141010.1 | - | 5 | 1222 | 9055622 |
