siRNA Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081394.1 | NP_001074863.1 | 7 | 2624 | ||
| NM_027854.1 | NP_082130.1 | 7 | 2686 | |||
| GenBank mRNA | AK171152.1 | - | 7 | 2653 | ||
| AK171607.1 | - | 4 | 1007 | |||
| BC033455.1 | - | 7 | 2594 | 4018584 | ||

Tmem248
Genomic Map