siRNA Details
| Gene Symbol: Ulk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027895.1 | NP_082171.1 | 16 | 2726 | ||
| GenBank mRNA | AK004783.1 | - | 14 | 3308 | ||
| AK032030.1 | - | 14 | 2348 | |||
| AK154642.1 | - | 16 | 2639 | |||
| BC037093.1 | - | 15 | 3053 | |||
| BC039958.1 | - | 12 | 2112 | |||

Ulk3
Genomic Map