siRNA Details
| Gene Symbol: Mgat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019239.2 | NP_062112.2 | 2 | 427 | ||
| XM_006242072.2 | XP_006242134.1 | 2 | 522 | |||
| XM_006242073.2 | XP_006242135.1 | 2 | 997 | |||
| XM_006242074.2 | XP_006242136.1 | 2 | 400 | |||
| XM_006242077.2 | XP_006242139.1 | 2 | 886 | |||
| GenBank mRNA | D10852.1 | - | 2 | 427 | ||

Mgat3
Genomic Map