siRNA Details
| Gene Symbol: Slc25a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081048.2 | NP_001074517.1 | 10 | 1343 | ||
| GenBank mRNA | AK005250.1 | - | 8 | 1255 | ||
| BC116828.1 | - | 10 | 1343 | 40126147 | ||
| BC137603.1 | - | 10 | 1343 | 8860622 | ||

Slc25a18
Genomic Map