siRNA Details
| Gene Symbol: 1700003E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027948.1 | NP_082224.1 | 4 | 655 | ||
| GenBank mRNA | AK005628.1 | - | 3 | 677 | ||
| BC126949.1 | - | 2 | 759 | 40090681 | ||
| CK137463.1 | - | 3 | 380 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700003E16Rik| Gene Symbol: 1700003E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027948.1 | NP_082224.1 | 4 | 655 | ||
| GenBank mRNA | AK005628.1 | - | 3 | 677 | ||
| BC126949.1 | - | 2 | 759 | 40090681 | ||
| CK137463.1 | - | 3 | 380 | |||