siRNA Details
| Gene Symbol: 1700003E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027948.1 | NP_082224.1 | 2 | 398 | ||
| GenBank mRNA | AK005628.1 | - | 2 | 338 | ||
| BC126949.1 | - | 2 | 301 | 40090681 | ||
| BF452482.1 | - | 2 | 300 | |||
| BY253424.1 | - | 2 | 399 | |||
| CK137463.1 | - | 1 | 123 | |||

1700003E16Rik
Genomic Map