siRNA Details
| Gene Symbol: Gpr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134385.2 | NP_001127857.1 | 4 | 2177 | ||
| NM_001134386.2 | NP_001127858.1 | 3 | 1660 | |||
| NM_001286994.1 | NP_001273923.1 | 4 | 2186 | |||
| NM_001286995.1 | NP_001273924.1 | 3 | 1889 | |||
| NM_001286996.1 | NP_001273925.1 | 4 | 1760 | |||
| NM_027965.2 | NP_082241.1 | 4 | 1989 | |||
| GenBank mRNA | AK154784.1 | - | 4 | 2144 | ||
| BY527936.1 | - | 1 | 371 | |||

Gpr160
Genomic Map