siRNA Details
| Gene Symbol: Cfap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027972.1 | NP_082248.1 | 2 | 123 | ||
| GenBank mRNA | AK006449.1 | - | 2 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap45| Gene Symbol: Cfap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027972.1 | NP_082248.1 | 2 | 123 | ||
| GenBank mRNA | AK006449.1 | - | 2 | 115 | ||