siRNA Details
| Gene Symbol: Cand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027994.1 | NP_082270.1 | 15 | 7559 | ||
| GenBank mRNA | AK038673.1 | - | 1 | 3215 | ||
| AK049323.1 | - | 1 | 1917 | |||
| AK054251.1 | - | 1 | 3470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cand1| Gene Symbol: Cand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027994.1 | NP_082270.1 | 15 | 7559 | ||
| GenBank mRNA | AK038673.1 | - | 1 | 3215 | ||
| AK049323.1 | - | 1 | 1917 | |||
| AK054251.1 | - | 1 | 3470 | |||