siRNA Details
| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027999.1 | NP_082275.1 | 14 | 1280 | ||
| GenBank mRNA | AK009476.1 | - | 14 | 1260 | ||
| AK129228.1 | - | 10 | 2548 | |||
| AK153920.1 | - | 14 | 1262 | |||
| BC023723.1 | - | 14 | 1230 | |||
| BC089002.1 | - | 14 | 1249 | |||
| BC118934.1 | - | 14 | 1255 | 40090953 | ||
| BC118935.1 | - | 14 | 1255 | 40090954 | ||

Haus5
Genomic Map